Peristaltic pumps, FlexiPump®
Product Details & Documents
FlexiPump® is a range of peristaltic pumps for culture media preparation. Use it for serial dispensing of culture media, agar or any liquid diluent for efficient sample preparation.
- Select from 3 dispensing modes: Continuous, dose (predetermined volume), multi-dose (predetermined volume, with an adjustable pause between each dose)
- 32 editable programs
- Rotating button for easy navigation
- Compact: Only 16 cm wide
- Precise flow control: Up to 1% precision
- Dispenses liquid from 50 μl to 99 L
- Contamination free: Dispensing assembly is entirely autoclavable with bottle
- Anti-drop system
- Easy dispensing
- Improved dispensing control with the handy gun and the adjustable arm
- Maintenance-free
- High quality Watson Marlow™ pump heads
Available in 2 models:
FlexiPump® with the essential functions
FlexiPump® Pro for high-performance dispensing
FlexiPump® is delivered with 1 peristaltic Watson Marlow™ pump head(s), 1 set of 3,2 mm dispensing assembly (nozzle + GL 45 bottle connector + filter + tubing +
tube weight), user's manual, power cord, free sterile packs of filter bags.
FlexiPump® Pro is delivered with 2 peristaltic Watson Marlow™ pump head(s), 1 set of 3,2 mm dispensing assembly (nozzle + GL 45 bottle connector + filter +
tubing + tube weight), 1 set of 6,4 mm dispensing assembly, user's manual, power cord, USB cable, free sterile packs of filter bags.
Certifications: In compliance with ISO 6887-1 and ISO 7218 standards
Spécifications
- Désignation:FlexiPump® Pack containing one FlexiPump® peristaltic pump (with single head and 3 rollers) and a handy gun for high precision work and confortable handling
- Seq:02
- Débit min.:1,5 ml/min
- Cat. No.:710-1888
- Ø intérieur tuyau:1,6 − 3,2 − 4,8 −6,4 − 8 mm
- Poids:5,2 kg
- Matériau du boîtier:Stainless steel
- Débit max.:1250 ml/min
- L×P×H:155×270×210 mm
- Vitesse de rotation:10−550 rpm
- Exigence électrique:100−240 V / 50−60 Hz
- Modèle:FlexiPump®
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- INSC561002Dispensing arm, adjustable, for use with other containersConnexion Pour la disponibilité1 * 1 ST - 374,00 €
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- 710-1887Handy gun, dispensing control with handy gun including electronic start buttonConnexion Pour la disponibilité1 * 1 ST - 362,00 €
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- INSC5130115 sets of silicone tubes for pump part (200 mm each), inner Ø 4,8 mmConnexion Pour la disponibilité1 * 1 SET - 78,00 €
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



