Handheld Digital Microscopes, Universal
Product Details & Documents
The Universal series provides a broad range of products with high image quality to satisfy the most demanding user. Magnification range: 10 to 220×.
- High speed USB 2.0 connection
- Metal or composite housing
- Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
- Wireless streaming to a tablet, Smart phone, or computer is possible in combination with WF-10 unit (to be ordered separately)
- Integrated polariser greatly reduces glare on reflective surfaces (some models only)
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED Control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Delivery information: Supplied with protecting pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)) and calibration target.
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : [email protected]
Documents
Dino-Lite Powerful Portable Versatile Dino-Lite Software
Product Literature
Specifications
- Grossissement:10 - 220×
- Modèle:UNIVERSAL, AM7915MZT
- Résolution:5,0 MP
- Désignation:Handheld microscope with exchangeable caps + polariser, Edge
- Seq:5
- Caractéristiques supplémentaires:AMR/EDOF/EDR/FLC
- Éclairage:8 LEDs
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- IDCPRK-06-PLExtension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)Log in for availability1 * 1 ST - 28,60 €
Product Information
- Catalog #DésignationAvailabilityPrice
- 630-1821Foot pedal, enables users to take a picture with their footLog in for availability1 * 1 ST - 63,30 €
Product Information
- Catalog #DésignationAvailabilityPrice
- 630-2517Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal armLog in for availability1 * 1 ST - 340,00 €
Product Information
- Catalog #Désignationl×p×hAvailabilityPrice
- IDCPRK-06ATabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX150×220×270 mmLog in for availability1 * 1 ST - 252,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
- 630-2923Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion batteryLog in for availability1 * 1 ST - 329,00 €
Product Information
- Catalog #Désignationl×p×hAvailabilityPrice
- IDCPRK-06AETabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EX150×220×270 mmLog in for availability1 * 1 ST - 249,00 €