Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Human PDGF BB ELISA Kits

Human PDGF BB ELISA Kits

Human PDGF BB ELISA kit, 96 tests
Catalog # ABCAAB184860-96
Supplier:   Abcam
Human PDGF BB ELISA Kits
Catalog # ABCAAB184860-96
Supplier:  Abcam
CAS Number:  
Human PDGF BB ELISA Kits
Human PDGF BB ELISA Kits

Human PDGF BB ELISA kit, 96 tests

CAS Number:  
Price
785,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human PDGF BB ELISA Kit is a single-wash 90-min Simplestep used to quantify Human PDGF BB with a sensitivity of 2,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.

Specifications

Specifications

  • Désignation:
    Human PDGF BB ELISA kit
  • Tests par set:
    96 tests
  • Type de test:
    Sandwich
  • Cat. No.:
    ABCAAB184860-96
  • Méthode de détection:
    Colorimetric
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Cible:
    PDGF B
  • Portée de détection:
    11,7 - 750 pg/ml
  • Type déchantillon:
    Plasma, cell culture media, serum
  • Sensibilité:
    2,9 pg/ml
  • Durée du test:
    One step
  • Time to Results:
    1 h 30 min
  • Température de stockage:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.