Cylinders, borosilicate glass, with hexagonal base
500 ml
SCOT213914478
DURAN® borosilicate glass 3.3, clear.
- Without graduation
- Hexagonal base
- Base with three knobs which increase its stability
: DIN EN ISO 4788.
- Capacité:500 ml
- Seq:07
DURAN® borosilicate glass 3.3, clear.
: DIN EN ISO 4788.