Cylinders, borosilicate glass, with hexagonal base
500 ml
  SCOT213914478
Cylinders, borosilicate glass, with hexagonal base
  SCOT213914478
Cylinders, borosilicate glass, with hexagonal base
Cylinders, borosilicate glass, with hexagonal base
500 ml
303,00 €
1 * 10 ST
DURAN® borosilicate glass 3.3, clear.
  • Without graduation
  • Hexagonal base
  • Base with three knobs which increase its stability
 : DIN EN ISO 4788.
  • Capacité:
    500 ml
  • Seq:
    07