Cylinders, borosilicate glass, with hexagonal base
10 ml
N° de catalogue SCOT213910879
Fournisseur: DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
N° de catalogue SCOT213910879
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
DURAN® borosilicate glass 3.3, clear.
- Without graduation
- Hexagonal base
- Base with three knobs which increase its stability
Certifications: DIN EN ISO 4788.
Spécifications
- Capacité:10 ml
- Seq:02