Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
β-Cyclodextrine, poudre blanche

β-Cyclodextrine, poudre blanche

5 g
N° de catalogue ICNA0219005305
Fournisseur:   MP Biomedicals
β-Cyclodextrine, poudre blanche
N° de catalogue ICNA0219005305
Fournisseur:  MP Biomedicals
Numéro CAS:  7585-39-9
β-Cyclodextrine, poudre blanche

5 g

Numéro CAS:  7585-39-9
Prix
33,00 €
Connexion Pour la disponibilité
1 * 5 g
Product Details & Documents

β-cyclodextrin is most commonly used as a complexing agent. It is reported to be useful for the selective precipitation of enantiomeric, positional or structural isomers. It also forms a water soluble complex with dansyl chloride for fluorescent labeling of proteins.

Cyclodextrins are cyclic oligosaccharides consisting of 6, 7, or 8 glucopyranose units with hydrophobic interiors, usually referred to as α-, β-, or γ-cyclodextrins, respectively. Lipophilic drugs of a size compatible with the hydrophobic core of a cyclodextrin can form complexes, resulting in increased aqueous solubility of the drugs. Cyclodextrins are non-toxic in many species (mice and rabbits), and do not denature proteins or interfere with enzymatic reactions.

Soluble in water, 1 M NH4OH: 50 mg/ml (clear, colorless solution).

Synonymes:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • Numéro CAS:7585-39-9
  • Formule:C₄₂H₇₀O₃₅
  • Numéro MDL:MFCD00078139
  • Poids moléculaire:1134.98 g/mol
  • Point de fusion:298…300 °C (lit.)
  • Densité:1,624 g/cm³ (lit.)
  • Température de stockage:Température ambiante
Spécifications

Résultats des tests de spécification

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
  • pH (1% aq soln):
    5 - 7.5
  • Loss on Drying:
    ≤14%
  • Optical Rotation:
    158 ± 6° (c=1,water)

Spécifications

  • Cdt:
    5 g

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.