Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
DEPC

DEPC

50 mL
N° de catalogue ICNA0215090250
Fournisseur:   MP Biomedicals
DEPC
N° de catalogue ICNA0215090250
Fournisseur:  MP Biomedicals
Numéro CAS:  1609-47-8
Numéro CAS:  1609-47-8
Prix
251,00 €
Connexion Pour la disponibilité
1 * 50 mL
Product Details & Documents

Diethyl Pyrocarbonate (DEPC) is a chemical use to inactivate RNase enzymes and is sensitive to moisture and pH. It is also sensitive to ammonia, which causes decomposition to urethane, a possible carcinogen.

It is effective as a nuclease inhibitor, modification reagent for His and Tyr residues in proteins. It is used as a robust probe for structural disruptions in dsDNA, reacting with fully or partially unstacked bases. It is used to modify proteins and nucleic acids. It is also used for Dot blot hybridisation, to dilute total RNA isolated from different micro-organisms.

Soluble in alcohols, esters, ketones, hydrocarbons. Slowly soluble in water with hydrolytic decomposition yielding ethanol and CO2.

Synonymes:  DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
  • Numéro CAS:1609-47-8
  • Formule:O(COOC₂H₅)₂
  • Numéro MDL:MFCD00009106
  • Poids moléculaire:162.14 g/mol
  • Température de stockage:Réfrigérateur
Spécifications

Spécifications

  • Cdt:
    50 mL

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.