Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Benzalkonium chlorure ≥80%, solution visqueuse

1 kg
N° de catalogue ICNA0215043091
Fournisseur:   MP Biomedicals
Benzalkonium chlorure ≥80%, solution visqueuse
N° de catalogue ICNA0215043091
Fournisseur:  MP Biomedicals
Numéro CAS:  8001-54-5
Benzalkonium chlorure ≥80%, solution visqueuse

1 kg

Numéro CAS:  8001-54-5
Prix
124,00 €
Connexion Pour la disponibilité
1 * 1 kg
Product Details & Documents

It has been reported that benzalkonium chloride inactivates HIV and HSV in vitro.

It is not effective against gram-negative bacteria or picornaviruses and it is not recommended for treatment of sanitising critical or semicritical medical instruments. Incompatible with anionic detergents, such as soap, and with nitrates. The preservative effect can be neutralized by the addition of Tween 80 and lecithin.

Soluble in water, alcohol, acetone; slightly soluble in benzene. Aqueous solutions are slightly alkaline to litmus and foams strongly when shaken. Composed of C12, C14, and C16 alkylbenzyldimethylammonium chlorides. Approximate ratios: C12 (40%) : C14 (50%) : C16 (10%).

 : Research Use Only (RUO).

Synonymes:  Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
  • Numéro CAS:8001-54-5
  • Formule:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
  • Numéro MDL:MFCD00145757
  • Température de stockage:Température ambiante
  • UN:2923
  • ADR:8,III
Spécifications

Résultats des tests de spécification

  • Identity Test:
    Passes
  • Concentration:
    80% Solution

Spécifications

  • Cdt:
    1 kg

Spécifications