1-Hexadécanol
Product Details & Documents
Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.
Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.
Easily soluble in diethyl ether, soluble in acetone, chloroform; slightly soluble in alcohol.
Store at +4 °C
- Numéro CAS:36653-82-4
- Formule:H₃C(CH₂)₁₄CH₂OH
- Numéro MDL:MFCD00004760
- Poids moléculaire:242.45 g/mol
- Index Merck:13,02037
- Point d'ébullition:305…330 °C (1013 hPa)
- Point de fusion:46…52 °C
- Densité:0,81 g/cm³ (20 °C)
- Point d'éclair:>110 °C
- Température de stockage:Température ambiante
- EINECS:253-149-0
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:100 g
Spécifications
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



