3,3',5,5'-Tétraméthylbenzidine ≥99%, en poudre
Product Details & Documents
3,3',5,5'-Tetramethylbenzidine is suitable as peroxidase substrate for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620 to 650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases. It is useful in gel staining procedure for low levels of home-associated peroxidases and for enzyme immunoassay of horseradish peroxidase.
Soluble in solvents 5%, Ethyl Acetate; insoluble in water. Dissolve 1,5 mg TMB in 0,1 ml of DMSO. Then add 9,9 ml of 0,1 M sodium acetate (pH 6,0). Finally, add hydrogen peroxide to a concentration of 0,01% and filter through a Whatman #1 paper filter. Prepare solutions fresh daily. Discard any remaining solution at the end of the day.
: Research Use Only (RUO).
- Numéro CAS:54827-17-7
- Formule:C₁₆H₂₀N₂
- Numéro MDL:MFCD00007748
- Poids moléculaire:240.35 g/mol
- Température de stockage:Température ambiante
Spécifications
Résultats des tests de spécification
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Melting Point:165 - 170 °C
Spécifications
- Cdt:5 g



