Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
3,3',5,5'-Tétraméthylbenzidine ≥99%, en poudre

3,3',5,5'-Tétraméthylbenzidine ≥99%, en poudre

100 mg
Catalog # ICNA02190277.1
Irritant
Supplier:   MP Biomedicals
3,3',5,5'-Tétraméthylbenzidine ≥99%, en poudre
Catalog # ICNA02190277.1
Supplier:  MP Biomedicals
CAS Number:  54827-17-7
3,3',5,5'-Tétraméthylbenzidine ≥99%, en poudre

100 mg

CAS Number:  54827-17-7
Price
27,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

3,3',5,5'-Tetramethylbenzidine is suitable as peroxidase substrate for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620 to 650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases. It is useful in gel staining procedure for low levels of home-associated peroxidases and for enzyme immunoassay of horseradish peroxidase.

Soluble in solvents 5%, Ethyl Acetate; insoluble in water. Dissolve 1,5 mg TMB in 0,1 ml of DMSO. Then add 9,9 ml of 0,1 M sodium acetate (pH 6,0). Finally, add hydrogen peroxide to a concentration of 0,01% and filter through a Whatman #1 paper filter. Prepare solutions fresh daily. Discard any remaining solution at the end of the day.

Caution: Research Use Only (RUO).

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Température ambiante
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • Melting Point:
    165 - 170 °C

Specifications

  • Cdt:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.