Embedding cassettes with disposable lid
rouge
N° de catalogue FTEC110-12-10LMI
Fournisseur: FA TECH DIAGNOSTICS
Embedding cassettes with disposable lid
N° de catalogue FTEC110-12-10LMI
Fournisseur: FA TECH DIAGNOSTICS
Aller à :
Product Details & Documents
Embedded cassettes with hinged lid.
- Cassettes maintain their stability, rigidness and shape during processing of tissues
: Available in different colours.
: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Spécifications
- Couleur:rouge
- Seq:86