Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
bleu clair
  FTEC110-08-10LMI
Embedding cassettes with disposable lid
  FTEC110-08-10LMI
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
bleu clair
101,00 €
1 * 1.000 ST
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
Ordering information: Available in different colours.
Packaging: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
  • Couleur:
    bleu clair
  • Seq:
    78