Microscope slides for printers
vert
KLINKP-SIL-3060
Microscope slides for slide printers.
- Frosted ends
- 90° ground edges
- Finished clipped corners of 1 mm
Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
- Couleur:vert
- Seq:12