Microscope slides for printers
fuchsia
N° de catalogue KLINKP-SIL-3059
Fournisseur: KLINIPATH
Aller à :
Product Details & Documents
Microscope slides for slide printers.
- Frosted ends
- 90° ground edges
- Finished clipped corners of 1 mm
Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
Spécifications
- Couleur:fuchsia
- Seq:11