Microscope slides for printers
Microscope slides for printers
jaune
Catalog # KLINKP-SIL-3058
Supplier:   KLINIPATH
Microscope slides for printers
Catalog # KLINKP-SIL-3058
Supplier:  KLINIPATH
Microscope slides for printers
jaune
Price
270,00 €
Log in for availability
1 * 2.500 ST
Product Details & Documents
Microscope slides for slide printers.
  • Frosted ends
  • 90° ground edges
  • Finished clipped corners of 1 mm
Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
Specifications
  • Couleur:
    jaune
  • Seq:
    10