Microscope slides for printers
Microscope slides
Catalog # 631-1229
Supplier: KLINIPATH
Jump to:
Product Details & Documents
Microscope slides for slide printers allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
- Frosted ends
- 90° grounded edges
- Finished clipped corners of 1mm
Specifications
- Désignation:Microscope slides
- Seq:01