Embedding cassettes with disposable lid
beige
N° de catalogue FTEC110-10-10LMI
Fournisseur:   FA TECH DIAGNOSTICS
Embedding cassettes with disposable lid
N° de catalogue FTEC110-10-10LMI
Fournisseur:  FA TECH DIAGNOSTICS
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
beige
Prix
128,00 €
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
 : Available in different colours.
 : Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Spécifications
  • Couleur:
    beige
  • Seq:
    82