Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
bleu
N° de catalogue KLIN110-03LM
Fournisseur:   KLINIPATH
Embedding cassettes with disposable lid
N° de catalogue KLIN110-03LM
Fournisseur:  KLINIPATH
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
bleu
Prix
217,00 €
Connexion Pour la disponibilité
1 * 1.600 ST
Product Details & Documents
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
Ordering information: Available in different colours.
Packaging: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Spécifications
  • Couleur:
    bleu
  • Seq:
    44