Accessoires
Male hose adapter, aluminum, DN 16 KF/16 mm int.Ø for PVC hose
Catalog # ILMV710052
Accessoires
Catalog # ILMV710052
Accessoires
Male hose adapter, aluminum, DN 16 KF/16 mm int.Ø for PVC hose
Price
22,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Désignation:
    Male hose adapter, aluminum, DN 16 KF/16 mm int.Ø for PVC hose
  • Seq:
    09