InfinityLab Stay safe caps
A-Line Stay Safe cap (GL45), 2-port, includes vent valve with time strip, 2 fittings (PFA) for Ø 3,2 mm tubing
HEWL5043-1218
: Agilent
InfinityLab Stay Safe caps stop evaporation of harmful solvent vapors from solvent bottles.
- Prevents 99,9% of solvents from leaching into the air
- Increase long-term mobile phase consistency
- GL 45 thread compatible with Agilent solvent bottles
These safety caps have been designed for optimal sealing with an integrated exhaust valve providing pressurisation during solvent extraction and allowing proper solvent flow to HPLC system. The exhaust valve contains a PTFE membrane to prevent contamination of your solvents from dirt and dust particles. Try the new A-Line Stay Safe caps for an airtight seal, complete with an innovative time strip that tells you when you need to change the valve or filter.
- Désignation:A-Line Stay Safe cap (GL45), 2-port, includes vent valve with time strip, 2 fittings (PFA) for Ø 3,2 mm tubing
Accessories
- Désignation
- Désignation
- Désignation
- Désignation
- Désignation
- HEWL5043-1221A-Line 6 L waste can kit. Includes A-Line Stay Safe cap (GL 45), 4-port for waste, PTFE waste collector, 2-port, must be used with a charcoal filter1 * 1 ST - 219,00 €
- Désignation
- HEWL5043-1789Solvent lines, in ultra clean tubing quality for bottle head caps, recommended for LC/MS application, 228 cm length, with premounted 1/4-28 PEEK fitting1 * 4 ST - 318,00 €
- Désignation
- HEWL5043-1190A-Line Stay Safe vent valve, PTFE, with time strip for A-Line Stay Safe cap (GL 45)1 * 1 ST - 31,00 €
- Désignation
- Désignation
- HEWL5043-1193A-Line Stay Safe charcoal filter, 58 g, with time strip for A-Line Stay Safe cap (GL 45), 4-port for waste (HEWL5043-1220)1 * 1 ST - 142,00 €
- Désignation
- Désignation
- Désignation
- HEWL5043-1790Solvent lines for bottle head caps, 228 cm length, with premounted 1/4-28 PEEK fitting1 * 4 ST - 54,00 €