VitraPOR® Micro-Filter Candle-Blanks
Product Details & Documents
VitraPOR® micro-filter candle blanks are ideal for integration into customised filter systems. Compact and chemically resistant, they provide reliable fine filtration in small volumes.
VitraPOR® porous filter
- Compact dimensions
- Ideal for custom assemblies
- High chemical and thermal resistance
- Suitable for fine filtration
- Customisable connection options
-
Reusable and durable
Material: Borosilicate glass 3.3
VitraPOR Micro-Filter Candle Blanks are designed for precision filtration tasks in compact or customised systems. Each blank features a VitraPOR glass filter element that ensures reliable and consistent particle retention. The borosilicate glass body offers excellent resistance to chemicals and high temperatures, making these blanks ideal for demanding laboratory and industrial applications. Whether for integration into housings, reactors, or special apparatus, ROBU candle blanks offer flexibility, robustness, and precise filtration performance.
Certifications: In compliance with ISO 4796-80 and ASTM E128-99 standards.
Documents
TDS VitraPOR Borosilicate Glass 3.3.pdf
Fiche technique
Specifications
- Ø×h:20×30 mm
- Porosité:2
- Désignation:Micro-filter candle-blank
- Seq:15