Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Moexipril Related Compounds, Reference Standard

Moexipril Related Compound C, USP reference standard, Vial, 20 mg
N° de catalogue 1445620.
Fournisseur:   USP
Moexipril Related Compounds, Reference Standard
N° de catalogue 1445620.
Fournisseur:  USP
Numéro CAS:  
Moexipril Related Compounds, Reference Standard

Moexipril Related Compound C, USP reference standard, Vial, 20 mg

Numéro CAS:  
Prix
509,00 €
Connexion Pour la disponibilité
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    20 mg
  • Normes:
    USP reference standard
  • Désignation:
    Moexipril Related Compound C
  • Cat. No.:
    1445620.
  • N ° CAS.:
    103733-40-0
  • Désignation:
    Moexipril Related Compound C
  • Synonyme(s):
    (S)-tert-butyl 2-{(S)-2-[(S)-1-ethoxy-1-oxo-4-phenylbutan-2-ylamino]propanoyl}-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, maleate

Spécifications