Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Diethyltoluamide, Reference Standards

Diethyltoluamide, Reference Standards

Diethyltoluamide Related Compound A, USP reference standard, Vial, 25 mg
N° de catalogue 1197018.
Fournisseur:   USP
Diethyltoluamide, Reference Standards
N° de catalogue 1197018.
Fournisseur:  USP
Numéro CAS:  
Diethyltoluamide, Reference Standards

Diethyltoluamide Related Compound A, USP reference standard, Vial, 25 mg

Numéro CAS:  
Prix
1 320,00 €
Connexion Pour la disponibilité
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    25 mg
  • Normes:
    USP reference standard
  • Désignation:
    Diethyltoluamide Related Compound A
  • Cat. No.:
    1197018.
  • N ° CAS.:
    2728-05-4
  • Formula:
    C₁₂H₁₇NO
  • Désignation:
    Diethyltoluamide Related Compound A
  • Synonyme(s):
    N,N-Diethyl-4-toluamide

Spécifications