Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Mobile storage cabinets

Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 1 door left, 450×516×740 mm
Catalog # KOTT315-00146
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00146
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 1 door left, 450×516×740 mm

Price
649,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Désignation:
    Mobile storage cabinet, with 1 drawer of 150 mm height
  • Seq:
    21
  • l×p×h:
    450×516×740 mm
  • Portes:
    1 door left
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.