Mobile storage cabinets
Mobile storage cabinet, With front open, 900×500×590 mm
N° de catalogue KOTT315-00128
Fournisseur: KOETTERMANN
Aller à :
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Spécifications
- Désignation:Mobile storage cabinet
- Seq:22
- l×p×h:900×500×590 mm
- Portes:With front open