Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 1 drawers, 450×516×740 mm
KOTT315-00121
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
- Désignation:Mobile storage cabinet, with 1 drawer of 150 mm height, lockable
- Seq:13
- l×p×h:450×516×740 mm
- Portes:1 drawers



