Mobile storage cabinets
Mobile storage cabinet, lockable, 1 door left, 450×516×740 mm
N° de catalogue KOTT315-00119
Fournisseur: KOETTERMANN
Aller à :
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Spécifications
- Désignation:Mobile storage cabinet, lockable
- Seq:12
- l×p×h:450×516×740 mm
- Portes:1 door left