Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Mobile storage cabinets

Mobile storage cabinets

Mobile storage cabinet, with 3 drawer of 150 mm height, pencil tray, lockable, 3 drawers, 450×516×590 mm
N° de catalogue KOTT315-00114
Fournisseur:   KOETTERMANN
Mobile storage cabinets
N° de catalogue KOTT315-00114
Fournisseur:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 3 drawer of 150 mm height, pencil tray, lockable, 3 drawers, 450×516×590 mm

Prix
903,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
Spécifications
  • Désignation:
    Mobile storage cabinet, with 3 drawer of 150 mm height, pencil tray, lockable
  • Seq:
    08
  • l×p×h:
    450×516×590 mm
  • Portes:
    3 drawers