Mobile storage cabinets
Mobile storage cabinet with 1 shelf, lockable, 1 door right, 450×516×590 mm
N° de catalogue KOTT315-00112
Fournisseur: KOETTERMANN
Aller à :
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Spécifications
- Désignation:Mobile storage cabinet with 1 shelf, lockable
- Seq:06
- l×p×h:450×516×590 mm
- Portes:1 door right