Mobile storage cabinets
Mobile storage cabinet, lockable, 2 doors, 1200×516×740 mm
N° de catalogue KOTT315-00101
Fournisseur: KOETTERMANN
Aller à :
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Spécifications
- Désignation:Mobile storage cabinet, lockable
- Seq:46
- l×p×h:1200×516×740 mm
- Portes:2 doors