Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Mobile storage cabinets

Mobile storage cabinets

Mobile storage cabinet, with 2 drawer of 150 mm height and 1 drawer of 300 mm height, lockable, 3 drawers, 1200×516×740 mm
N° de catalogue KOTT315-00098
Fournisseur:   KOETTERMANN
Mobile storage cabinets
N° de catalogue KOTT315-00098
Fournisseur:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 2 drawer of 150 mm height and 1 drawer of 300 mm height, lockable, 3 drawers, 1200×516×740 mm

Prix
1 350,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
Spécifications
  • Désignation:
    Mobile storage cabinet, with 2 drawer of 150 mm height and 1 drawer of 300 mm height, lockable
  • Seq:
    43
  • l×p×h:
    1200×516×740 mm
  • Portes:
    3 drawers