Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 1200×500×590 mm
N° de catalogue KOTT315-00092
Fournisseur: KOETTERMANN
Aller à :
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Spécifications
- Désignation:Mobile storage cabinet with 1 shelf
- Seq:37
- l×p×h:1200×500×590 mm
- Portes:With front open