General purpose gloves, High performance, A700
, XL, noir
N° de catalogue PRTWA700BKRXL
Fournisseur: Portwest
Aller à :
Product Details & Documents
This high-performing, hand-hugging glove is multi-purpose and suitable for any job.
- Reinforced stitching on the thumb for extra strength
- Low linting construction for minimal contamination
- Wrist support and protection
- Wrist strap for secure fitting
Lightweight for the wearer, this glove has durability built in and features reinforced stitching on the thumb for extra strength with wrist support and protection.
Suitable for mechanic applications, general construction, material handling, carpentry and utility work.
: EN 388:2016 (1111X)
Spécifications
- Taille du gant:XL
- Couleur:noir
- Seq:03
- Cat. No.:PRTWA700BKRXL
- Usage unique/Réutilisable:réutilisable
- Norme EN:EN 388:2016 (1111X)
- Ambidextre/anatomique:Conception main droite et main gauche
- Modèle:A700
- Style de manchette:À Velcro®
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



