Reagent troughs
Disposable reagent trough for maximum reagent recovery, Tecan Pure, 25 ml, gris
N° de catalogue TCAN30055743
Marketsource
Fournisseur: Tecan
Aller à :
Product Details & Documents
These disposable PP troughs are designed to optimise the performance and productivity of Tecan liquid handling platforms.
Spécifications
- Capacité:25 ml
- Couleur:gris
- Désignation:Disposable reagent trough for maximum reagent recovery, Tecan Pure
- Seq:01