Inserts
E 143, insert 1/4, mesh tray, for various items, with 2 hinged handles, 5 mm mesh spacing, 5 mm frame all round, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 255×230×55 mm
MIEL3830310
: MIELE
- Désignation:E 143, insert 1/4, mesh tray, for various items, with 2 hinged handles, 5 mm mesh spacing, 5 mm frame all round
- Seq:7
- l×p×h:255×230×55 mm
- Pour:PG 8535/36, PG 8582, PG 8582 CD, PG 8592