Jars, with screw cap
250 ml, 65 mm, 83 mm
Catalog # SPVAPIPXX0250
Supplier: GRAVIS
Jump to:
Product Details & Documents
PP, white.
Specifications
- Capacité:250 ml
- Hauteur:83 mm
- Seq:01
- Ø col:65 mm