Phase separating filters
Phase separating filters
108 H, 240 mm
N° de catalogue PRATA240108H
Fournisseur:   PRAT DUMAS
Phase separating filters
N° de catalogue PRATA240108H
Fournisseur:  PRAT DUMAS
Phase separating filters
Phase separating filters
108 H, 240 mm
Prix
207,00 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Delivery information: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
Spécifications
  • N°:
    108 H
  • Seq:
    06
  • Taille:
    240 mm