Phase separating filters
Phase separating filters
108 H, 185 mm
N° de catalogue PRATA185108H
Fournisseur:   PRAT DUMAS
Phase separating filters
N° de catalogue PRATA185108H
Fournisseur:  PRAT DUMAS
Phase separating filters
Phase separating filters
108 H, 185 mm
Prix
137,00 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Delivery information: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
Spécifications
  • N°:
    108 H
  • Seq:
    04
  • Taille:
    185 mm