Phase separating filters
Phase separating filters
108 H, 150 mm
N° de catalogue PRATA150108H
Fournisseur:   PRAT DUMAS
Phase separating filters
N° de catalogue PRATA150108H
Fournisseur:  PRAT DUMAS
Phase separating filters
Phase separating filters
108 H, 150 mm
Prix
101,00 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Livraison: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
Spécifications
  • N°:
    108 H
  • Seq:
    04
  • Taille:
    150 mm