Bottle racks
Bottle rack, 6× 1 litre (max. 100 mm diameter)
N° de catalogue HEIP1465250001
Fournisseur: MERCK LIFE SCIENCE
Aller à :
Product Details & Documents
Racks for media bottles, made of high quality stainless steel, electrolytic polished.
- Autoclavable
- Easy to clean
- Good chemical and temperature resistance
Safe and convenient transport of bottles. Suitable for isolators and clean rooms.
Spécifications
- Désignation:Bottle rack, 6× 1 litre (max. 100 mm diameter)
- Seq:02