Accessoires pour epMotion®
Accessoires pour epMotion®
Rack for 24 Safe-Lock-Tubes without temperature control, 0,5/1,5/2,0 ml, for use with reaction vessels, glass or plastic vessels of various sizes
N° de catalogue EPPE5075751.453
Fournisseur:   EPPENDORF
Accessoires pour epMotion®
N° de catalogue EPPE5075751.453
Fournisseur:  EPPENDORF
Accessoires pour epMotion®
Accessoires pour epMotion®
Rack for 24 Safe-Lock-Tubes without temperature control, 0,5/1,5/2,0 ml, for use with reaction vessels, glass or plastic vessels of various sizes
Prix
715,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spécifications
  • Désignation:
    Rack for 24 Safe-Lock-Tubes without temperature control, 0,5/1,5/2,0 ml, for use with reaction vessels, glass or plastic vessels of various sizes
  • Seq:
    32