Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Chromatography columns with filter plates

Chromatography column with PTFE tap, 20,00 mm, 200 mm, 29/32 NS
  NEUB1202290200200
Chromatography columns with filter plates
  NEUB1202290200200
Chromatography columns with filter plates

Chromatography column with PTFE tap, 20,00 mm, 200 mm, 29/32 NS

 
204,00 €
   
1 * 1 ST
 

These chromatography columns feature a top sleeve with PTFE tap and are made from borosilicate glass 3.3.

 
  • Ø int.:
    20,00 mm
  • Rodage femelle:
    29/32 NS
  • Désignation:
    Chromatography column with PTFE tap
  • Longueur:
    200 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.