Chromatography data station, DataApex ClarityVA
Clarity Chromatography Data Station upgrade to Clarity VA with Hitachi Chromaster and Primaide™ drivers included
N° de catalogue 906-0190
Fournisseur: COLLECTION PREFIX HPLC
Chromatography data station, DataApex ClarityVA
N° de catalogue 906-0190
Fournisseur: COLLECTION PREFIX HPLC
Aller à :
Product Details & Documents
ClarityVA is a flexible and powerful Chromatography Data System (CDS) that controls HPLC and GC systems. This modular software includes a lot of controls for many different systems, modules and interfaces. The flexibility enables an easy control of HPLC systems in combination with a user friendly surface.
- Direct control of Hitachi HPLC (Chromaster and Primaide)
- Modules for specific calculation and methodology
- Tools for regulated environment
- Extended user support
- Language localisations (English, German, French,...)
ClarityVA is useable for modern complained pharma labs, when configured correctly (21 CFR part 11 compliance).
Spécifications
- Désignation:Clarity Chromatography Data Station upgrade to Clarity VA with Hitachi Chromaster and Primaide™ drivers included
- Seq:05