Chromatography data station, DataApex ClarityVA
Chromatography data station, DataApex ClarityVA
Clarity VA Chromatography Data Station Offline version for data processing with Hitachi Chromaster and Primaide™ drivers included
N° de catalogue 906-0189
Fournisseur:   COLLECTION PREFIX HPLC
Chromatography data station, DataApex ClarityVA
N° de catalogue 906-0189
Fournisseur:  COLLECTION PREFIX HPLC
Chromatography data station, DataApex ClarityVA
Chromatography data station, DataApex ClarityVA
Clarity VA Chromatography Data Station Offline version for data processing with Hitachi Chromaster and Primaide™ drivers included
Prix
425,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
ClarityVA is a flexible and powerful Chromatography Data System (CDS) that controls HPLC and GC systems. This modular software includes a lot of controls for many different systems, modules and interfaces. The flexibility enables an easy control of HPLC systems in combination with a user friendly surface.
  • Direct control of Hitachi HPLC (Chromaster and Primaide)
  • Modules for specific calculation and methodology
  • Tools for regulated environment
  • Extended user support
  • Language localisations (English, German, French,...)
ClarityVA is useable for modern complained pharma labs, when configured correctly (21 CFR part 11 compliance).
Spécifications
  • Désignation:
    Clarity VA Chromatography Data Station Offline version for data processing with Hitachi Chromaster and Primaide™ drivers included
  • Seq:
    14