Electron multipliers
Electron multipliers
Electron multiplier for Agilent 7900 ICP-MS
N° de catalogue HEWLG8400-60258
Fournisseur:   Agilent
Electron multipliers
N° de catalogue HEWLG8400-60258
Fournisseur:  Agilent
Electron multipliers
Electron multipliers
Electron multiplier for Agilent 7900 ICP-MS
Prix
4 360,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The electron multiplier converts incident ions into an amplified electric current used to record measurements of the ion signal in ICP-MS Systems.
As the voltage in the system increases the amplification factor (or gain) increases. In general the electron multiplier can measure a count of 1.6E10. The system should be replaced when the total ion count reaches more than 3.2E11 and the sensitivity is less than 4000 counts with 10 ppb Y (even when applying detector voltage above 2700 V).
Spécifications
  • Désignation:
    Electron multiplier for Agilent 7900 ICP-MS
  • Seq:
    02
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.