Spray chambers and brackets
Spray chambers and brackets
Double-pass spray chamber, glass cyclonic design with ball joint socket and UniFit drain outlet, for use with organic solvents, high matrix or high TDS samples with 5100 ICP-OES
N° de catalogue HEWLG8010-60256
Fournisseur:   Agilent
Spray chambers and brackets
N° de catalogue HEWLG8010-60256
Fournisseur:  Agilent
Spray chambers and brackets
Spray chambers and brackets
Double-pass spray chamber, glass cyclonic design with ball joint socket and UniFit drain outlet, for use with organic solvents, high matrix or high TDS samples with 5100 ICP-OES
Prix
1 180,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Spécifications
  • Désignation:
    Double-pass spray chamber, glass cyclonic design with ball joint socket and UniFit drain outlet, for use with organic solvents, high matrix or high TDS samples with 5100 ICP-OES
  • Seq:
    04