Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
R-Phycoerythrin (RPE)

R-Phycoerythrin (RPE)

R-Phycoerythrin (RPE), SureLight® R-PE dye, 565>498 nm/578 nm, 1 mg
N° de catalogue COBSD5-000
Fournisseur:   Columbia Biosciences
R-Phycoerythrin (RPE)
N° de catalogue COBSD5-000
Fournisseur:  Columbia Biosciences
Numéro CAS:  
R-Phycoerythrin (RPE)
R-Phycoerythrin (RPE)

R-Phycoerythrin (RPE), SureLight® R-PE dye, 565>498 nm/578 nm, 1 mg

Numéro CAS:  
Prix
22,30 €
Connexion Pour la disponibilité
1 * 1 mg
Product Details & Documents

Many applications and instruments were developed specifically for R-phycoerythrin.

It is commonly used in immunoassays such as FACS, flow cytometry, multimer/tetramer applications. With new instrumetation available, R-PE is also well suited for immunohistochemistry (IHC) and Western blot applications.

Spécifications

Spécifications

  • Cdt:
    1 mg
  • Désignation:
    R-Phycoerythrin (RPE), SureLight® R-PE dye
  • Excitation/Émission:
    565>498 nm/578 nm
  • Cat. No.:
    COBSD5-000
  • Application:
    Flow cytometry, Cell-based assays, microarrays and microplate applications
  • Température de stockage:
    Store at 4 °C

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.