Cartridge filters, Optimizer® DEV and DI
Product Details & Documents
These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.
- Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, end caps, cage and core: HDPE.
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 2,7 bar at 25 °C
Maximum reverse differential pressure: 2,7 bar at 25 °C
Spécifications
- Surface de filtration:1600 cm²
- Taille de pore:0,1 µm
- Type:DEV 16 cartridge filter with 2-015 connections, Kalrez® O-rings, Ø×L: 67×52 mm
- Seq:03
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



