Cartridge filters, Optimizer® DEV and DI
These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.
- Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, end caps, cage and core: HDPE.
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 2,7 bar at 25 °C
Maximum reverse differential pressure: 2,7 bar at 25 °C
- Surface de filtration:3200 cm²
- Taille de pore:0,05 µm
- Type:DI 40 cartridge filter with 2-118 connections, Kalrez® O-rings, Ø×L: 67×123 mm
- Seq:12



